Order Entry
Denmark
ContactUsLinkComponent
6-(Fluorescein-5-carboxamido) hexanoic acid succinimidyl ester (5-SFX) ≥95% (by TLC) amine-reactive fluorescent dye
6-(Fluorescein-5-carboxamido) hexanoic acid succinimidyl ester (5-SFX) ≥95% (by TLC) amine-reactive fluorescent dye
Catalog # 90095.
Supplier:  Biotium
CAS Number:  
undefined
6-(Fluorescein-5-carboxamido) hexanoic acid succinimidyl ester (5-SFX) ≥95% (by TLC) amine-reactive fluorescent dye
Catalog # 90095.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :586.55 g/mol
Storage Temperature:Freezer
 :148356-00-7
Formula:C₃₁H₂₆N₂O₁₀