Order Entry
Denmark
ContactUsLinkComponent
6-Carboxyfluorescein-N-succinimidyl ester (6-FAM SE) ≥99% (by TLC) amine-reactive fluorescent dye
6-Carboxyfluorescein-N-succinimidyl ester (6-FAM SE) ≥99% (by TLC) amine-reactive fluorescent dye
  90023.
 :  Biotium
 :  
undefined
6-Carboxyfluorescein-N-succinimidyl ester (6-FAM SE) ≥99% (by TLC) amine-reactive fluorescent dye
  90023.
 :  Biotium
 :  
  • Pk:
    10 mg
  • Seq:
    2275
 :
 :473.39 g/mol
 :Freezer
 :92557-81-8
 :C₂₅H₁₅NO₉
 :MFCD00077323