Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Ordreindtastning
Ordrestatus
Din konto
Kurv
Denmark
Ordreindtastning
Ordrestatus
Din konto
Kurv
ContactUsLinkComponent
Home
Standards
Organic Standards
Pharmaceutical secondary standard, Chloramphenicol, (CRM), Supelco®
Pharmaceutical secondary standard, Chloramphenicol, (CRM), Supelco®
Pharmaceutical secondary standard, Chloramphenicol, (CRM), Supelco®
Catalog # PHR1412-1G
Supplier:
Merck
CAS Number:
Udvid billedet
Pharmaceutical secondary standard, Chloramphenicol, (CRM), Supelco®
Catalog # PHR1412-1G
Supplier:
Merck
CAS Number:
Specifications
Pk:
1 g
CAS No.:
56-75-7
Description:
Chloramphenicol
Description:
Chloramphenicol
Specifications
About this item
Product Details
Product Details