0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
ContactUsLinkComponent
Standards
Organic Standards
Pharmaceutical secondary standard, Cetyl Palmitate, (CRM), Supelco®
Pharmaceutical secondary standard, Cetyl Palmitate, (CRM), Supelco®
PHR1166-1G
:
Merck
:
Pharmaceutical secondary standard, Cetyl Palmitate, (CRM), Supelco®
PHR1166-1G
:
Merck
:
Pk:
1 g
CAS No.:
540-10-3
Description:
Cetyl Palmitate
Description:
Cetyl Palmitate