Order Entry
Denmark
ContactUsLinkComponent
Pharmaceutical secondary standard, ascorbic acid, (CRM), Supelco®
Pharmaceutical secondary standard, ascorbic acid, (CRM), Supelco®
Supplier:  Merck
Certificates
undefined
Pharmaceutical secondary standard, ascorbic acid, (CRM), Supelco®
Supplier:  Merck

Specifications

  • CAS No.:
    50-81-7
  • Melting point:
    190…192 °C
  • Formula:
    C₆H₈O₆
  • Description:
    Ascorbic acid

Specifications

Related Information

Frequently Bought Together