Supelco® TraceCERT® Organic Reference Standard, Acetophenone
63634-50MG
: Merck
:
- Pk:50 mg
- Standards:Certified reference material, TraceCERT®
- CAS No.:98-86-2
- Description:Acetophenone
- Storage conditions:15...25 °C
- Hazard class description:H302 - H319
- Shipping conditions:STD
- MDL No.:MFCD00008724
- M-Clarity quality level:MQ300
- Synonym(s):Methyl phenyl ketone
- Melting point:19 - 20 °C (lit.)
- Formula:CH₃COC₆H₅
- Molecular weight:120,15
- Density:1,03 g/ml at 25 °C (lit.)
- Flash point:82 °C - Closed cup
- Boiling point:202 °C (lit.)
- GHS signal: GHS07
: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
: P264 - P280 - P301 + P312 - P305 + P351 + P338 - P337 + P313 - P501