Order Entry
Denmark
ContactUsLinkComponent
Supelco® Cerilliant® Organic Reference Standard, Nicotinamide-¹³C₆ solution
Supelco® Cerilliant® Organic Reference Standard, Nicotinamide-¹³C₆ solution
Catalog # V-034-1ML
Supplier:  Merck
CAS Number:  
Supelco® Cerilliant® Organic Reference Standard, Nicotinamide-¹³C₆ solution
Catalog # V-034-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml in methanol
  • Pk:
    1 mL
  • Standards:
    Certified reference material, Cerilliant®
  • Description:
    Nicotinamide-¹³C₆ solution
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H370
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    Vitamin B3-¹³C₆ solution
  • Molecular weight:
    128,08
  • Flash point:
    9,7 °C - Closed cup
  • Pk Info:
    Ampule of 1 ml

Specifications

About this item

  • GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311