Supelco® Drug and metabolite standards, Epitestosterone solution, (CRM)
E-058-1ML
: Merck
:
- Concentration:1,0 mg/ml in acetonitrile
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:481-30-1
- Description:Epitestosterone solution
- Environmentally Preferable:
- Description:Epitestosterone solution
- Pk Info:ampoule of 1 ml