Order Entry
Denmark
ContactUsLinkComponent
Supelco® Drug and metabolite standards, Atropine solution, (CRM)
Supelco® Drug and metabolite standards, Atropine solution, (CRM)
Catalog # A-046-1ML
Supplier:  Merck
CAS Number:  
Supelco® Drug and metabolite standards, Atropine solution, (CRM)
Catalog # A-046-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1,0 mg/ml in acetonitrile
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    51-55-8
  • Description:
    Atropine solution
  • Description:
    Atropine solution
  • Pk Info:
    ampoule of 1 ml

Specifications

About this item