Supelco® Cerilliant® Organic Reference Standard, 3-Methoxytyramine hydrochloride
Catalog # M-171-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:1,0 mg/ml in methanol (as free base)
- Pk:1 mL
- Standards:Certified reference material, Cerilliant®
- CAS No.:1477-68-5
- Description:3-Methoxytyramine hydrochloride
- Storage conditions:–20 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Shipping conditions:DICE
- MDL No.:MFCD00012902
- M-Clarity quality level:MQ300
- Synonym(s):3-MT; 3-Methoxy-4-hydroxyphenethylamine; 4-(2-Aminoethyl)-2-methoxyphenol hydrochloride
- Formula:CH₃OC₆H₃-4-(OH)CH₂CH₂NH₂·HCl
- Molecular weight:203,67
- Flash point:9,7 °C - Closed cup
Specifications
About this item
- GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311