Order Entry
Denmark
ContactUsLinkComponent
Industrial gloves, uvex profi ergo XG20A
Industrial gloves, uvex profi ergo XG20A
Supplier:  UVEX
Certificates
undefined
Industrial gloves, uvex profi ergo XG20A
Supplier:  UVEX
Specifications
  • Catalog No:
  • UVEX6055810
  • UVEX6055808
  • UVEX6055809
  • UVEX6055807
  • Model:
  • XG20A
  • XG20A
  • XG20A
  • XG20A
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Cotton
  • Cotton
  • Cotton
  • Cotton
  • Finish:
  • Special NBR and Xtra Grip coating on palm, fingers and ¾ of the back
  • Special NBR and Xtra Grip coating on palm, fingers and ¾ of the back
  • Special NBR and Xtra Grip coating on palm, fingers and ¾ of the back
  • Special NBR and Xtra Grip coating on palm, fingers and ¾ of the back
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (3121X)
  • EN 388:2016 (3121X)
  • EN 388:2016 (3121X)
  • EN 388:2016 (3121X)