Cut resistant gloves, uvex unidur 6643
: UVEX
These sensitive cut protection gloves with HPPE fibres and NBR coating on palm and fingertips are lightweight and provide a cut protection level of 3/B. They are also extremely sensitive.
- Very high abrasion, tear and puncture resistance
- Good grip in damp and oily areas
- High flexibility and good tactile feel
- Good wearer comfort
Ideal for use in the automotive and sheet metal industry, assembly, repair and maintenance work.
: EN 388:2016 (4344B)
- Catalog No:
- UVEX6031408
- UVEX6031407
- UVEX6031410
- UVEX6031409
- Model:
- 6643
- 6643
- 6643
- 6643
- Ambidextrous/Hånd-specifik:
- Hånd-specifik
- Hånd-specifik
- Hånd-specifik
- Hånd-specifik
- Engangs/Genbrugelig:
- Genanvendelig
- Genanvendelig
- Genanvendelig
- Genanvendelig
- Foringsmateriale:
- HPPE / polyamid / elastan
- HPPE / polyamid / elastan
- HPPE / polyamid / elastan
- HPPE / polyamid / elastan
- Overflade:
- NBR coating on palm and fingertips
- NBR coating on palm and fingertips
- NBR coating on palm and fingertips
- NBR coating on palm and fingertips
- Manchet:
- Strikket
- Strikket
- Strikket
- Strikket
- EN norm:
- EN 388:2016 (4344B)
- EN 388:2016 (4344B)
- EN 388:2016 (4344B)
- EN 388:2016 (4344B)
Frequently Bought Together