Thermostatic baths for viscosity measurement, TC-650
Supplier: AMETEK
Certificates
About this item
The compact TC-650 is a circulating refrigerated bath for use with water-jacketed devices such as cone plate, small sample adapter, UL adapter accessory, RST-CC Plus rheometer and RST-CPS Plus rheometer.
- Provides standalone operation - no tap water required
- Easy control of set-point
- Accommodates one 600 ml beaker
Programmable controller version is designed to automate sample temperature control.
Frequently Bought Together