- Model:DV2TRV
- No. of speeds:200
- Range:100** - 40 M mPa•s
- Revolutions:0,1 - 200 min-1
- Seq:02
- Pk:1
These versatile viscometers features a 5-inch colour display to guide users through test creation and data gathering for fast and easy viscosity measurements.
- Built-in RTD temperature probe
- Internal data storage: 150 MB
- Enhanced security with customisable user levels & password access to comply with regulatory requirements such as 21 CFR Part 11
- Built-In options: Timed tests, data averaging, programmable QC limits/alarms, customizable speed/spindle lists, on screen data comparison
- Auto range shows maximum viscosity measured with any spindle/speed combination
- USB PC interface provides optional computer control and automatic data gathering capability
- Download custom test programs with PG Flash Software (included with instrument)
Displayed information:
- Viscosity (cP or mPa•s)
- Temperature (°C or °F)
- Shear Rate/Stress
- % Torque
- Speed/Spindle
- Step Program Status
Each viscometer is supplied with an internal spring used to calculate the viscosity of a material. These springs have different measuring capabilities:
LV is for low viscosity materials and can measure the thinnest materials. Typical examples include inks, oils, and solvents.
RV is for medium viscosity materials. Typical examples include creams, food, and paints.
* 1 cP achieved with UL Adapter accessory. 15 cP on LV with standard spindles.
** Minimum viscosity is achieved with optional RV/HA/HB spindle.