Chemical resistant gloves, u-chem 3300
: UVEX
These highly sensitive chemical protection gloves with gauntlet, made of seamless bamboo-viscose fibre and a nitrile rubber coating, are kind to skin and comfortable to wear even for long periods.
- Extremely lightweight and highly flexible
- Very good fit
- Good resistance to many chemicals
- Silicone-free according to imprint test
- Low-linting
Ideal for use in chemical industry, printing industry, maintenance and inspection work, laboratory tasks, painting work, food processing, pharmaceutical industry and in cleaning.
: EN 388:2016 (2121X), EN ISO 374-1:2016/Type A (JKLOPT).
- Catalog No:
- 112-5412
- 112-5413
- 112-5416
- Model:
- 3300
- 3300
- 3300
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Liner material:
- Nylon
- Nylon
- Nylon
- Finish:
- Fully coated
- Fully coated
- Fully coated
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet
- EN norm:
- EN 388:2016 (2121X), EN ISO 374-1:2016 Type A (JKLOPT)
- EN 388:2016 (2121X), EN ISO 374-1:2016 Type A (JKLOPT)
- EN 388:2016 (2121X), EN ISO 374-1:2016 Type A (JKLOPT)
- Food contact approved:
- +
- +
- +
Frequently Bought Together