Order Entry
Denmark
ContactUsLinkComponent
Trichoderma viride cellulase
Trichoderma viride cellulase
Supplier:  US Biological
Certificates
Trichoderma viride cellulase
Supplier:  US Biological

Specifications

  • CAS No.:
    9012-54-8
  • Species:
    Trichoderma viride
  • Form:
    Light yellow solid
  • Enzyme name:
    Cellulase
  • pH:
    4,0 - 5,0 (optimal pH)

Specifications

Frequently Bought Together