Order Entry
Denmark
ContactUsLinkComponent
Reporter gene assays, SpectraMax® DuoLuc™ reporter assay kit
Reporter gene assays, SpectraMax® DuoLuc™ reporter assay kit
  MLDVR8361
 :  
undefined
Reporter gene assays, SpectraMax® DuoLuc™ reporter assay kit
  MLDVR8361
 :  
  • Pk:
    1 KIT
  • Description:
    SpectraMax® DuoLuc™ Reporter Assay kit (Explorer), 200 reactions
SpectraMax® DuoLuc™ reporter assay kit offers sensitive detection of dual luciferase reporter gene assays, which are important tools for monitoring gene expression and other cellular events.
  • Enables highly sensitive quantitation of both firefly and Renilla luciferases in mammalian cells
  • Flash-type luminescence with excellent sensitivity
  • Expression measurement in as few as ten cells per well
  • Improved accuracy of results by normalising reporter expression to a control
  • Optimised for SpectraMax® i3x, iD3, or L microplate readers with injectors
  • Simplified data acquisition and analysis with preconfigured protocol in SoftMax® Pro Software