Order Entry
Denmark
ContactUsLinkComponent
Ramsbottom coking bulb
Ramsbottom coking bulb
Catalog # 201-1419
Supplier:  DWK Life Sciences
undefined
Ramsbottom coking bulb
Catalog # 201-1419
Supplier:  DWK Life Sciences
Specifications
  • Description:
    Ramsbottom bulb, O.Ø×H: 25×57 mm
  • Pk:
    25
Specifications
About this item
Ramsbottom coking bulbs are used in the determination of carbon residue in petroleum products.
Certifications: ASTM D524. Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.