DNA transfection kit for protein production, FectoCHO® Expression system
Supplier: Polyplus-transfection
Certificates
About this item
FectoCHO® expression system consists in the synergistic association of a novel CHO chemically defined medium, FectoCHO® CD expression medium and a powerful transfection reagent, FectoPRO®.
- Outstanding protein production yields in various CHO cells
- Fast and effortless CHO cell adaptation
- Chemically defined kit components
- Excellent lot to lot reproducibility
- Cost-effective solution
FectoCHO® CD expression medium allows easy cultivation of various strains of CHO cells such as CHO-K1, CHO-S and ExpiCHO®-S , without the need of an extensive sequential adaptation. FectoPRO®-mediated transient transfection in this chemically defined medium leads to the highest protein and antibody yields ever reached using Transient Gene Expression, up to 4 times higher than with similar systems.
Certifications: Polyplus-transfection® has been an ISO 9001 accredited company since 2002. Since then this certification has been successfully renewed every three years.
Ordering information: FectoPRO® transfection reagent and FectoCHO® CD expression medium can be purchased separately.
Specifications
- Bioproduction:Yes
- Cell specific:HEK-293, CHO
- DNA and siRNA:Yes
- DNA:Yes
- siRNA:Yes