Order Entry
Denmark
ContactUsLinkComponent
dNTP mix, 2 mM
dNTP mix, 2 mM
Catalog # R0242
CAS Number:  
undefined
dNTP mix, 2 mM
Catalog # R0242
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    dNTP mix, 2 mM
Specifications
About this item
dNTP mix contains premixed aqueous solutions of dATP, dCTP, dGTP and dTTP, each at a final concentration of 2 mM.
  • Greater than 99% purity confirmed by HPLC
  • Free of human and E. coli DNA
  • Highly stable
Suitable for used in all molecular biology applications, including PCR, real-time PCR, high fidelity and long PCR, LAMP-PCR, cDNA synthesis, RT-PCR, RDA, MDA, DNA labelling, and DNA sequencing.