Order Entry
Denmark
Orders LinkContactUsLinkComponent
Analytical funnel

Analytical funnel

Analytical funnel, white, sterilised, 47 mm, 0,45 µm
Catalog # APPCAFW-045MC
Supplier:   PanReac AppliChem
Analytical funnel
Catalog # APPCAFW-045MC
Supplier:  PanReac AppliChem
Analytical funnel
Analytical funnel

Analytical funnel, white, sterilised, 47 mm, 0,45 µm

Price
2.890,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

This sterilised analytical funnel is individually packed.

Specifications
  • Description:
    Analytical funnel, white, sterilised, 47 mm, 0,45 µm