Astaxanthine ≥97%, crystalline solid
Supplier: Cayman Chemical
Certificates
About this item
CAS Number:472-61-7
:
Formula:C₄₀H₅₂O₄
MDL Number:MFCD00672621
:596.852 g/mol
Storage Temperature:Freezer
Specification Test Results
- Purity:≥97%
- λmax:473 nm
Specification Test Results
Frequently Bought Together