Leukotriene B₄ Express ELISA kit
Supplier: Cayman Chemical
Certificates
About this item
A competitive assay that can be used for quantification of LTB₄ in plasma and other sample matrices.
- Assay 24 samples in triplicate or 36 samples in duplicate
- Measure LTB₄ levels down to 45 pg/ml
- Rapid assay; get results in under five hours
LTB₄ stimulates a number of leukocyte functions, including aggregation, stimulation of ion fluxes, enhancement of lysosomal enzyme release, superoxide anion production, chemotaxis, and chemokinesis.
LTB₄ is metabolized in leukocytes and hepatocytes to less active 20-hydroxy and 20-carboxy LTB₄ and then further oxidized to ω-carboxy dinor LTB₄ and ω-carboxy tetranor-LTB3. It is not excreted in the urine.
Delivery: Kit contains antibody, tracer, standard, ELISA and wash buffer concentrate, polysorbate 20, plate and cover, ellman's reagent, tracer and antiserum dye.
Specifications
- Mid point:100 - 200 pg/ml (50% B/B₀)
- Detection range:15,6 - 2000 pg/ml
- Storage temperature:–20 °C
- Sensitivity:45 pg/ml (80% B/B₀)
Specifications
Frequently Bought Together