Order Entry
Denmark
ContactUsLinkComponent
Echinocandin B
Echinocandin B
Supplier:  Cayman Chemical
Certificates
undefined
Echinocandin B
Supplier:  Cayman Chemical

Specifications

  • Storage conditions:
    Freezer
  • Form:
    Powder
  • Protein/peptide name:
    Echinocandin B
  • Purity:
    >95%
  • CAS No.:
    54651-05-7

Specifications

Related Information

Frequently Bought Together