Order Entry
Denmark
ContactUsLinkComponent
 
8061A Surrogate standard (3 components)
Katalog # REST31848
Leverandør:  Restek
CAS nummer:  
undefined
8061A Surrogate standard (3 components)
Katalog # REST31848
Leverandør:  Restek
CAS nummer:  

Specifikationer

  • Concentration:
    500 μg/ml each in acetone
  • Pk:
    1 Ampoul
  • Description:
    CLP 04.1 BNA surrogate mix
  • Pk Info:
    1 ml/ampoule

Specifikationer

Om denne vare

The standard consists of dibenzyl phthalate, diphenyl isophthalate and diphenyl phthalate.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.