Order Entry
Denmark
ContactUsLinkComponent
DRO/ORO calibration standard
DRO/ORO calibration standard
Catalog # REST31831
Supplier:  Restek
CAS Number:  
undefined
DRO/ORO calibration standard
Catalog # REST31831
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    25000 µg/ml each in methylene chloride
  • Pk:
    1 Ampoul
  • Description:
    DRO/ORO calibration standard
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

This consists of 10W30 motor oil:diesel fuel #2 (1:1 blend).