Order Entry
Denmark
ContactUsLinkComponent
Acetate mix (5 components)
  REST30477
 :  Restek
 :  
undefined
Acetate mix (5 components)
  REST30477
 :  Restek
 :  
  • Concentration:
    2000 µg/ml each in P and T methanol
  • Pk:
    1 mL
  • Description:
    8260 Acetate Mix
  • Pk Info:
    1 ml/ampoule
This contains: n-butyl acetate; ethyl acetate; isopropyl acetate; n-propyl acetate; vinyl acetate.