Order Entry
Denmark
ContactUsLinkComponent
ASTM D3606 calibration standard #3 without internal standard (2 components)
ASTM D3606 calibration standard #3 without internal standard (2 components)
Catalog # REST30649
Supplier:  Restek
CAS Number:  
undefined
ASTM D3606 calibration standard #3 without internal standard (2 components)
Catalog # REST30649
Supplier:  Restek
CAS Number:  

Specifications

  • Pk:
    1 Ampoul
  • Description:
    ASTM D3606 calibration standard #3 without internal standard, in isooctane
  • Pk Info:
    25 ml/ampoule

Specifications

About this item

The mix consists of benzene (1,25% vol/vol), toluene (10% vol/vol).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.