Order Entry
Denmark
ContactUsLinkComponent
ASTM® D3606 calibration standard #1 without internal standard (2 components)
ASTM® D3606 calibration standard #1 without internal standard (2 components)
Catalog # REST30647
Supplier:  Restek
CAS Number:  
undefined
ASTM® D3606 calibration standard #1 without internal standard (2 components)
Catalog # REST30647
Supplier:  Restek
CAS Number:  

Specifications

  • Pk:
    1 Ampoul
  • Description:
    ASTM D3606 calibration standard #1 without internal standard, in isooctane
  • Pk Info:
    25 ml/ampoule

Specifications

About this item

The mix consists of benzene (5% vol/vol), toluene (20% vol/vol).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.