Order Entry
Denmark
ContactUsLinkComponent
Method 525.2 GC-MS performance check mix, (3 components)
Method 525.2 GC-MS performance check mix, (3 components)
Catalog # REST31827
Supplier:  Restek
CAS Number:  
undefined
Method 525.2 GC-MS performance check mix, (3 components)
Catalog # REST31827
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml each in acetone
  • Pk:
    1 Ampoul
  • Description:
    Method 525.2 GC-MS performance check mix
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

This consists of 4,4'-DDT, DFTPP (decafl uorotriphenylphosphine) and endrin.