Order Entry
Denmark
ContactUsLinkComponent
 
Chlorinated acid herbicide mix (2 components)
Catalog # REST32429
Supplier:  Restek
CAS Number:  
undefined
Chlorinated acid herbicide mix (2 components)
Catalog # REST32429
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    1000 μg/ml each in acetonitrile
  • Pk:
    1 mL
  • Description:
    Chlorinated acid herbicide mix
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Contains two components, those are 2,4-dichlorophenoxyacetic acid and 2,4,5-TP.

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.