MA EPH matrix spike mix (10 components)
Catalog # REST31460
Supplier: Restek
CAS Number:
Specifications
- Concentration:250 µg/ml each in acetone
- Pk:1 mL
- Description:MA EPH matrix spike mix
- Pk Info:1 ml/ampoule
Specifications
About this item
Contains: n-nonane, C9, n-tetradecane, C14, n-nonadecane, C19, n-eicosane, C20, n-octacosane, C28, acenaphthene, anthracene, chrysene, naphthalene , pyrene.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.