Order Entry
Denmark
ContactUsLinkComponent
Method 525.2 PCB congener mix (8 components)
Method 525.2 PCB congener mix (8 components)
Catalog # REST32420
Supplier:  Restek
CAS Number:  
undefined
Method 525.2 PCB congener mix (8 components)
Catalog # REST32420
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    200 μg/ml each in acetone
  • Pk:
    1 mL
  • Description:
    Method 525.2 PCB congener mix
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Contains eight components, those are 2-chlorobiphenyl, 2,3-dichlorobiphenyl, 2,4,5-trichlorobiphenyl, 2,2',4,4'-tetrachlorobiphenyl, 2,2',3',4,6-pentachlorobiphenyl, 2,2',4,4',5,6'-hexachlorobiphenyl, 2,2',3,3',4,4',6-heptachlorobiphenyl and 2,2',3,3',4,5',6,6'-octachlorobiphenyl.

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.