Order Entry
Denmark
ContactUsLinkComponent
Organophosphorus pesticide mix (16 components), european formulation
Organophosphorus pesticide mix (16 components), european formulation
Catalog # REST32418
Supplier:  Restek
CAS Number:  
undefined
Organophosphorus pesticide mix (16 components), european formulation
Catalog # REST32418
Supplier:  Restek
CAS Number:  
Specifications
  • Pk:
    1 mL
  • Description:
    Organophosphorus pesticide mix, european formulation, in acetone
  • Environmentally Preferable:
  • Pk Info:
    1 ml/ampoule
Specifications
About this item
Contains 16 components, those are acephate, azinphos methyl, chlorpyrifos, demeton-s-methyl, dichlorvos, dimethoate, ethion, malathion, methamidophos, methidathion, omethoate, pirimiphos methyl, profenofos, prothiofos, pyrazophos and tolclofos-methyl.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.