Order Entry
Denmark
ContactUsLinkComponent
524 Calibration mix #8 (12 components)
524 Calibration mix #8 (12 components)
  REST30203
 :  Restek
 :  
undefined
524 Calibration mix #8 (12 components)
  REST30203
 :  Restek
 :  
  • Concentration:
    2000 μg/ml each in P and T methanol
  • Pk:
    1
  • Description:
    Calibration mix #8
  • Pk Info:
    1 ml/ampoule
Mix Contains: Carbon disulphide (75-15-0); Chloroacetonitrile (107-14-2); 1-Chlorobutane (Butyl chloride) (109-69-3); trans-1,4-Dichloro-2-butene (110-57-6); 1,1-Dichloro-2-propanone (513-88-2); Diethyl ether (ethyl ether) (60-29-7); Hexachloroethane (67-72-1); Iodomethane (methyl iodide) (74-88-4); Methacrylonitrile (126-98-7); Methyl tert-butyl ether (MTBE) (1634-04-4); 2-Nitropropane (79-46-9); Propionitrile (107-12-0).
 : Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.