Ordreindtastning
Denmark
ContactUsLinkComponent
Calibration mix #7 Kit, 524
Calibration mix #7 Kit, 524
Catalog # REST30202
Supplier:  Restek
CAS Number:  
undefined
Calibration mix #7 Kit, 524
Catalog # REST30202
Supplier:  Restek
CAS Number:  

Specifications

  • Koncentration:
    Contains 1 ml each of these mixtures
  • Pakn:
    1
  • Beskrivelse:
    Calibration mix #7 kit

Specifications

About this item

Contains 1 ml each of these mixtures: 524 calibration mix #7A, 524 calibration mix #7B. calibration mix #7A contains Acetone (67-64-1), 2-Butanone (MEK) (78-93-3), 2-Hexanone (591-78-6), 4-Methyl-2-pentanone (MIBK) (108-10-1), Tetrahydrofuran (109-99-9); calibration mix #7B contains Acrylonitrile (107-13-1)
Allyl chloride (3-chloropropene) (107-05-1), Ethyl methacrylate (97-63-2), Methyl acrylate (96-33-3), Methyl methacrylate (80-62-6), Nitrobenzene (98-95-3), Pentachloroethane (76-01-7).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.