Order Entry
Denmark
ContactUsLinkComponent
Calibration check mix, 8270 B/N (7 components)
Calibration check mix, 8270 B/N (7 components)
Catalog # REST31616
Supplier:  Restek
CAS Number:  
undefined
Calibration check mix, 8270 B/N (7 components)
Catalog # REST31616
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml each in methylene chloride
  • Pk:
    1 mL
  • Description:
    Calibration check mix, 8270 B/N
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Contains: Acenaphthene, benzo(a)pyrene, 1,4-dichlorobenzene, di-n-octyl phthalate, diphenylamine, fluoranthene, hexachlorobutadiene.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.