Residual solvents class 2 mix B (8 components)
Catalog # REST36272
Supplier: Restek
CAS Number:
Specifications
- Concentration:In dimethyl sulfoxide
- Pk:1 Ampoul
- Description:Residual solvents class 2 mix B
- Pk Info:1 ml/ampoule
Specifications
About this item
Contains: n-hexane, C6, 1450 µg/ml, chloroform, 300 µg/ml, 1,2-dimethoxyethane, 500 µg/ml, 2-hexanone, 250 µg/ml, nitromethane, 250 µg/ml, pyridine, 1000 µg/ml, tetralin, 500 µg/ml, trichloroethene, 400 µg/ml.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.