Grob DA 280 column test mix (11 components)
Catalog # REST35252
Supplier: Restek
CAS Number:
Specifications
- Concentration:In methylene chloride
- Pk:1
- Description:Grob DA 280 column test mix
- Pk Info:1 ml/ampoule
Specifications
About this item
Contains: n-decane, C10, 280 µg/ml, methyl decanoate, C10:0, 420 µg/ml, n-undecane, C11, 280 µg/ml, methyl undecanoate, C11:0, 420 µg/ml, methyl dodecanoate, C12:0, 420 µg/ml, L(+)-2,3-butanediol, 530 µg/ml, 2,6-dimethylaniline, 320 µg/ml, 2,6-dimethylphenol, 320 µg/ml, 2-ethylhexanoic acid, 380 µg/ml, nonanal, 400 µg/ml, 1-octanol, 360 µg/ml.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.