Canadian drinking water phenoxyacid herbicides mix (11 components)
Catalog # REST31868
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 μg/ml each in acetone
- Pk:1 Ampoul
- Description:Canadian drinking water phenoxyacid herbicides mix
- Pk Info:1 ml/ampoule
Specifications
About this item
Consists of 11 components, those are bromoxynil, 2,4-d, dicamba, 2,4-dichlorophenol, diclofop methyl, dinoseb, pentachlorophenol, picloram, 2,4,5-t, 2,3,4,6-tetrachlorophenol and 2,4,6-trichlorophenol.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.