Order Entry
Denmark
ContactUsLinkComponent
Pesticide standard mix A (9 components)
Pesticide standard mix A (9 components)
Catalog # REST32297
Supplier:  Restek
CAS Number:  
undefined
Pesticide standard mix A (9 components)
Catalog # REST32297
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    In hexane:toluene (90:10)
  • Pk:
    1 mL
  • Description:
    Pesticide standard mix A
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Consists of nine components, those are α-bhc, γ-bhc, 4,4'-ddd, 4,4'-ddt, dieldrin, endosulfan i, endrin, heptachlor and methoxychlor.

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.