Order Entry
Denmark
ContactUsLinkComponent
Pesticides mix #2, method 527 (5 components)
Pesticides mix #2, method 527 (5 components)
Catalog # REST33008
Supplier:  Restek
CAS Number:  
undefined
Pesticides mix #2, method 527 (5 components)
Catalog # REST33008
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    500 µg/ml each in ethyl acetate
  • Pk:
    1 Ampoul
  • Description:
    Pesticides mixture #2, method 527
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

The pesticides mix consists of five components chlorpyrifos, dimethoate, malathion, parathion and terbufos sulphone.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.