Order Entry
Denmark
ContactUsLinkComponent
USP  calibration mixture #5 (5 components)
USP  calibration mixture #5 (5 components)
Catalog # REST36007
Supplier:  Restek
CAS Number:  
undefined
USP  calibration mixture #5 (5 components)
Catalog # REST36007
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    In dimethyl sulphoxide
  • Pk:
    1 mL
  • Description:
    USP <467> calibration mixture #5
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

The mixture consists of benzene (2 μg/ml), chloroform (60 μg/ml), 1,4-dioxane (380 μg/ml), methylene chloride (dichloromethane) (600 μg/ml) and trichloroethene (80 μg/ml).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.