Order Entry
Denmark
ContactUsLinkComponent
LC reversed phase test mix #1 (4 components)
LC reversed phase test mix #1 (4 components)
Catalog # REST35005
Supplier:  Restek
CAS Number:  
undefined
LC reversed phase test mix #1 (4 components)
Catalog # REST35005
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    In methanol:water (75:25)
  • Pk:
    1 mL
  • Description:
    LC reversed phase test mixture #1
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Routine analysis using this mix can assist in determining the need to perform column and/or system maintenance.

The LC reversed phase test mix #1 consists of four components, they are benzene (3,00 mg/ml), biphenyl (0.06 mg/ml), naphthalene (0,50 mg/ml) and uracil (0,02 mg/ml).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.