Order Entry
Denmark
ContactUsLinkComponent
 
NJDEP EPH 10/08 rev. 2 aromatics matrix spike mix (18 components)
Catalog # REST30543
Supplier:  Restek
CAS Number:  
undefined
NJDEP EPH 10/08 rev. 2 aromatics matrix spike mix (18 components)
Catalog # REST30543
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    200 µg/mL each in acetone:toluene (50:50)
  • Pk:
    5 mL
  • Description:
    NJDEP EPH 10/08 rev.2 aromatics matrix spike mix
  • Pk Info:
    5 ml/ampoule

Specifications

About this item

Consists of 18 components, those are acenaphthene, acenaphthylene, anthracene, benzanthracene, benzopyrene, benzofluoranthene, benzoperylene, benzofluoranthene, chrysene, dibenzanthracene, fluoranthene, fluorene, indenopyrene, phenanthrene, 1,2,3-trimethylbenzene and pyrene.

18 components

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.