Order Entry
Denmark
ContactUsLinkComponent
SV calibration mix #4 (13 components)
SV calibration mix #4 (13 components)
Catalog # REST31010
Supplier:  Restek
CAS Number:  
undefined
SV calibration mix #4 (13 components)
Catalog # REST31010
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 μg/ml each in methylene chloride
  • Pk:
    1 mL
  • Description:
    SV calibration mix #4
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

SV Calibration mix #4 contains carbazole (86-74-8); dibenzofuran (132-64-9); diethyl phthalate (84-66-2); 2,4-dinitrotoluene (121-14-2); 2,6-dinitrotoluene (606-20-2); hexachlorobenzene (118-74-1); hexachlorobutadiene (87-68-3); hexachlorocyclopentadiene (77-47-4); hexachloroethane (67-72-1); isophorone (78-59-1); 2-methylnaphthalene (91-57-6); nitrobenzene (98-95-3); 1,2,4-trichlorobenzene (120-82-1).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.