Order Entry
Denmark
ContactUsLinkComponent
GC multiresidue pesticide standard #4 (28 components)
GC multiresidue pesticide standard #4 (28 components)
Catalog # REST32566
Supplier:  Restek
CAS Number:  
undefined
GC multiresidue pesticide standard #4 (28 components)
Catalog # REST32566
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml each in toluene
  • Pk:
    1 mL
  • Description:
    GC multiresidue pesticide standard #4
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Organonitrogen compounds that has accurately identify and quantify pesticide residues by GC-MS/MS in fruits, vegetables, botanicals, and herbals such as tea, ginseng, ginger, echinacea, and dietary supplements.

Contains: Acetochlor, alachlor, allidochlor, clomazone (command), cycloate, diallate(cis & trans), dimethachlor, diphenamid, fenpropathrin, fluquinconazole, flutolanil, linuron, metazachlor, methoxychlor, metolachlor, N-(2,4-dimethylphenyl)formamide, norflurazon, oxadiazon, pebulate, pretilachlor, prochloraz, propachlor, propanil, propisochlor, propyzamide, pyridaben, tebufenpyrad, triallate.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.