Order Entry
Denmark
ContactUsLinkComponent
GC multiresidue pesticide standard #3 (25 components)
GC multiresidue pesticide standard #3 (25 components)
Catalog # REST32565
Supplier:  Restek
CAS Number:  
undefined
GC multiresidue pesticide standard #3 (25 components)
Catalog # REST32565
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml each in toluene:acetonitrile (99:1)
  • Pk:
    1 mL
  • Description:
    GC multiresidue pesticide standard #3
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Organonitrogen Compounds that has accurately identify and quantify pesticide residues by GC-MS/MS in fruits, vegetables, botanicals, and herbals such as tea, ginseng, ginger, echinacea, and dietary supplements.

Contains: Benfluralin, biphenyl, chlorothalonil, dichlofluanid, dichloran, 3,4-dichloroaniline, 2,6-dichlorobenzonitrile (dichlobenil), diphenylamine, ethalfluralin, fluchloralin, isopropalin, nitralin, nitrofen, oxyfluorfen, pendimethalin, pentachloroaniline, pentachlorobenzonitrile, pentachloronitrobenzene (quintozene), prodiamine, profluralin, 2,3,5,6-tetrachloroaniline, tetrachloronitrobenzene (tecnazene), THPI (tetrahydrophthalimide), tolylfluanid, trifluralin.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.