Order Entry
Denmark
ContactUsLinkComponent
 
Extraction solvent stock solution #2 (2 components)
Catalog # REST31680
Supplier:  Restek
CAS Number:  
Extraction solvent stock solution #2 (2 components)
Catalog # REST31680
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    In hexane
  • Pk:
    1
  • Description:
    Extraction Solvent Stock Solution #2
  • Pk Info:
    20 ml/ampoule

Specifications

About this item

This extraction solvent stock solution contains: n-decane (C10) (20 µl/l), n-tetracontane (C40) (20 mg/l).