Order Entry
Denmark
ContactUsLinkComponent
Extraction solvent stock solution #1 (2 components)
Extraction solvent stock solution #1 (2 components)
Catalog # REST31679
Supplier:  Restek
CAS Number:  
undefined
Extraction solvent stock solution #1 (2 components)
Catalog # REST31679
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    In hexane, 5 ml/ampoule
  • Pk:
    1
  • Description:
    Extraction Solvent Stock Solution #1

Specifications

About this item

This extraction solvent stock solution contains: n-decane (C10), n-tetracontane (C40).